Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTATCAGGAATGTCAAATGACTC[C/T]TCTTCAAAATCATCATCCTTCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607385 | ||||||||||||||||||||
Literature Links: |
MGAT4C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MGAT4C - MGAT4 family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013244.3 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | NP_037376.2 | |
XM_005268776.4 | 1328 | Silent Mutation | GAA,GAG | E,E 361 | XP_005268833.1 | |
XM_005268779.3 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_005268836.1 | |
XM_011538150.2 | 1328 | Silent Mutation | GAA,GAG | E,E 370 | XP_011536452.1 | |
XM_011538151.2 | 1328 | Silent Mutation | GAA,GAG | E,E 370 | XP_011536453.1 | |
XM_011538156.2 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_011536458.1 | |
XM_011538157.2 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_011536459.1 | |
XM_017019143.1 | 1328 | Silent Mutation | GAA,GAG | E,E 381 | XP_016874632.1 | |
XM_017019144.1 | 1328 | Silent Mutation | GAA,GAG | E,E 370 | XP_016874633.1 | |
XM_017019145.1 | 1328 | Silent Mutation | GAA,GAG | E,E 361 | XP_016874634.1 | |
XM_017019146.1 | 1328 | Silent Mutation | GAA,GAG | E,E 361 | XP_016874635.1 | |
XM_017019147.1 | 1328 | Silent Mutation | GAA,GAG | E,E 361 | XP_016874636.1 | |
XM_017019148.1 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_016874637.1 | |
XM_017019149.1 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_016874638.1 | |
XM_017019150.1 | 1328 | Silent Mutation | GAA,GAG | E,E 332 | XP_016874639.1 |