Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGACTGGTTCTTCAGTACCTC[C/T]ACCAGGAAGGAATCTTGACGGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRADD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRADD - CASP2 and RIPK1 domain containing adaptor with death domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320099.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | NP_001307028.1 | |
NM_001320100.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | NP_001307029.1 | |
NM_001320101.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | NP_001307030.1 | |
NM_003805.4 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | NP_003796.1 | |
XM_017020142.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016875631.1 | |
XM_017020143.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016875632.1 | |
XM_017020144.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016875633.1 | |
XM_017020145.1 | 222 | Missense Mutation | CAC,TAC | H,Y 31 | XP_016875634.1 |