Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAACTGCTGGGGCAGGTCGTGGG[C/T]GAGGAGGTCCCCTGCTGTAGGGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603437 | ||||||||||||||||||||
Literature Links: |
YBX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YBX3 - Y-box binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145426.1 | Intron | NP_001138898.1 | ||||
NM_003651.4 | Intron | NP_003642.3 | ||||
XM_011520871.2 | Intron | XP_011519173.1 | ||||
XM_017020122.1 | Intron | XP_016875611.1 | ||||
XM_017020123.1 | Intron | XP_016875612.1 |