Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCCTCTCCATCTCCCCCAGTT[A/G]AAGAAGAACAGGAGAAAAGACTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC63 - coiled-coil domain containing 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286243.1 | 149 | UTR 5 | NP_001273172.1 | |||
NM_001286244.1 | 149 | Intron | NP_001273173.1 | |||
NM_152591.2 | 149 | Silent Mutation | TTA,TTG | L,L 4 | NP_689804.1 | |
XM_006719262.1 | 149 | Silent Mutation | TTA,TTG | L,L 4 | XP_006719325.1 | |
XM_006719263.2 | 149 | Silent Mutation | TTA,TTG | L,L 4 | XP_006719326.1 | |
XM_011537999.1 | 149 | Silent Mutation | TTA,TTG | L,L 4 | XP_011536301.1 | |
XM_011538001.2 | 149 | Silent Mutation | TTA,TTG | L,L 4 | XP_011536303.1 |