Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGTGACTGTAAGCTGCCTCTGC[A/G]TGGCATCCAGCACAGCCCGGTACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601478 | ||||||||||||||||||||
Literature Links: |
MYO1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO1A - myosin IA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256041.1 | 3223 | Missense Mutation | ACG,ATG | T,M 991 | NP_001242970.1 | |
NM_005379.3 | 3223 | Missense Mutation | ACG,ATG | T,M 991 | NP_005370.1 | |
XM_011538373.2 | 3223 | Intron | XP_011536675.1 |