Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGTAATTTTAAGAATGGCTTTT[C/G]AACAAATGTGGTAGAGGCAGGTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM186A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM186A - family with sequence similarity 186 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145475.1 | 6921 | Missense Mutation | CAA,GAA | Q,E 2262 | NP_001138947.1 | |
XM_006719231.3 | 6921 | Missense Mutation | CAA,GAA | Q,E 2262 | XP_006719294.1 | |
XM_011537890.2 | 6921 | Missense Mutation | CAA,GAA | Q,E 2258 | XP_011536192.1 | |
XM_011537892.1 | 6921 | Missense Mutation | CAA,GAA | Q,E 2119 | XP_011536194.1 |