Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGAGAAGGCGCGGCGCGCTCTGG[A/G]CGCCCTGGCCGCTGCCCTGAGGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164350 MIM: 603351 | ||||||||||||||||||||
Literature Links: |
OAS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OAS1 - 2'-5'-oligoadenylate synthetase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OAS3 - 2'-5'-oligoadenylate synthetase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006187.3 | 120 | Missense Mutation | GAC,GGC | D,G 34 | NP_006178.2 | |
XM_005253889.3 | 120 | Missense Mutation | GAC,GGC | D,G 34 | XP_005253946.1 | |
XM_017019363.1 | 120 | Missense Mutation | GAC,GGC | D,G 34 | XP_016874852.1 |