Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGAGATCCGCTTCTACCAGCT[G/T]GGGGACGAGGCCCTGGCGGCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176257 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNA6 - potassium voltage-gated channel subfamily A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002235.3 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | NP_002226.1 | |
XM_005253686.3 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | XP_005253743.1 | |
XM_011520955.2 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | XP_011519257.1 | |
XM_017019270.1 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | XP_016874759.1 | |
XM_017019271.1 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | XP_016874760.1 | |
XM_017019272.1 | 1244 | Silent Mutation | CTG,CTT | L,L 126 | XP_016874761.1 |