Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAATCCAGTCAATTATAATGAG[C/T]GCCATGCTTCCAATCATGAAGAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608065 | ||||||||||||||||||||
Literature Links: |
SLC38A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC38A4 - solute carrier family 38 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143824.1 | 1992 | Silent Mutation | GCA,GCG | A,A 531 | NP_001137296.1 | |
NM_018018.4 | 1992 | Silent Mutation | GCA,GCG | A,A 531 | NP_060488.2 | |
XM_005268997.2 | 1992 | Silent Mutation | GCA,GCG | A,A 531 | XP_005269054.1 |