Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCCTCAGCTCCATCACTGTCT[C/T]GGGCTGAAAGCCAAAGGAGGGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM234B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM234B - family with sequence similarity 234 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020853.1 | 3339 | Intron | NP_065904.1 | |||
XM_017019706.1 | 3339 | Intron | XP_016875195.1 |
GPRC5D - G protein-coupled receptor class C group 5 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018654.1 | 3339 | Missense Mutation | CAA,CGA | Q,R 300 | NP_061124.1 | |
XM_005253421.3 | 3339 | Missense Mutation | CAA,CGA | Q,R 300 | XP_005253478.1 | |
XM_017019582.1 | 3339 | Missense Mutation | CAA,CGA | Q,R 300 | XP_016875071.1 | |
XM_017019583.1 | 3339 | Intron | XP_016875072.1 |