Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGAGCGAAGTCCATGATGCT[C/T]GGGTCGAGGGCATCCACCTCAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXN4 - forkhead box N4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213596.2 | 1563 | Silent Mutation | CCA,CCG | P,P 423 | NP_998761.2 | |
XM_011537922.2 | 1563 | Silent Mutation | CCA,CCG | P,P 424 | XP_011536224.1 | |
XM_011537923.2 | 1563 | Intron | XP_011536225.1 | |||
XM_017018818.1 | 1563 | Silent Mutation | CCA,CCG | P,P 380 | XP_016874307.1 | |
XM_017018819.1 | 1563 | Intron | XP_016874308.1 |