Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTATGGAGAGGGCAGGACCAGCC[A/G]GGGAGGAGGGCGGCGCGCGCGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605453 MIM: 607668 MIM: 608920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB9 - ATP binding cassette subfamily B member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARL6IP4 - ADP ribosylation factor like GTPase 6 interacting protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002251.2 | 350 | Missense Mutation | AGG,GGG | R,G 69 | NP_001002251.2 | |
NM_001002252.2 | 350 | Missense Mutation | AGG,GGG | R,G 69 | NP_001002252.2 | |
NM_001278378.1 | 350 | Missense Mutation | AGG,GGG | R,G 69 | NP_001265307.1 | |
NM_001278379.1 | 350 | Intron | NP_001265308.1 | |||
NM_001278380.1 | 350 | UTR 5 | NP_001265309.1 | |||
NM_016638.3 | 350 | Missense Mutation | AGG,GGG | R,G 69 | NP_057722.3 | |
NM_018694.3 | 350 | Missense Mutation | AGG,GGG | R,G 69 | NP_061164.3 |
OGFOD2 - 2-oxoglutarate and iron dependent oxygenase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304833.1 | 350 | Intron | NP_001291762.1 | |||
NM_001304834.1 | 350 | Intron | NP_001291763.1 | |||
NM_001304835.1 | 350 | Intron | NP_001291764.1 | |||
NM_001304836.1 | 350 | Intron | NP_001291765.1 | |||
NM_001304837.1 | 350 | Intron | NP_001291766.1 | |||
NM_001304838.1 | 350 | Intron | NP_001291767.1 | |||
NM_024623.2 | 350 | Intron | NP_078899.1 |
PITPNM2 - phosphatidylinositol transfer protein membrane associated 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |