Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGGAGAAGATGAAGGAGGAGAA[A/G]AGGAGGAGTCTGGAGGCCCGACGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612654 | ||||||||||||||||||||
Literature Links: |
TCHP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCHP - trichoplein keratin filament binding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143852.1 | 405 | Silent Mutation | AAA,AAG | K,K 76 | NP_001137324.1 | |
NM_032300.4 | 405 | Silent Mutation | AAA,AAG | K,K 76 | NP_115676.1 | |
XM_011538836.2 | 405 | Silent Mutation | AAA,AAG | K,K 76 | XP_011537138.1 | |
XM_011538837.2 | 405 | Silent Mutation | AAA,AAG | K,K 76 | XP_011537139.1 | |
XM_017020024.1 | 405 | Silent Mutation | AAA,AAG | K,K 76 | XP_016875513.1 | |
XM_017020025.1 | 405 | UTR 5 | XP_016875514.1 |