Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGCCCCTGGTGTGGCAGACTTGG[C/T]CTTGTCTGAGAACTGGGCCCAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600414 | ||||||||||||||||||||
Literature Links: |
PEX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEX5 - peroxisomal biogenesis factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000319.4 | 695 | Missense Mutation | GCC,GTC | A,V 113 | NP_000310.2 | |
NM_001131023.1 | 695 | Missense Mutation | GCC,GTC | A,V 128 | NP_001124495.1 | |
NM_001131024.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | NP_001124496.1 | |
NM_001131025.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | NP_001124497.1 | |
NM_001131026.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | NP_001124498.1 | |
NM_001300789.1 | 695 | Missense Mutation | GCC,GTC | A,V 134 | NP_001287718.1 | |
XM_005253455.1 | 695 | Missense Mutation | GCC,GTC | A,V 134 | XP_005253512.1 | |
XM_011520793.2 | 695 | Missense Mutation | GCC,GTC | A,V 149 | XP_011519095.1 | |
XM_011520795.1 | 695 | Missense Mutation | GCC,GTC | A,V 128 | XP_011519097.1 | |
XM_011520797.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_011519099.1 | |
XM_011520798.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_011519100.1 | |
XM_011520799.2 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_011519101.1 | |
XM_011520800.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_011519102.1 | |
XM_011520801.2 | 695 | Missense Mutation | GCC,GTC | A,V 149 | XP_011519103.1 | |
XM_011520802.2 | 695 | Missense Mutation | GCC,GTC | A,V 30 | XP_011519104.1 | |
XM_017019744.1 | 695 | Missense Mutation | GCC,GTC | A,V 261 | XP_016875233.1 | |
XM_017019745.1 | 695 | Missense Mutation | GCC,GTC | A,V 261 | XP_016875234.1 | |
XM_017019746.1 | 695 | Missense Mutation | GCC,GTC | A,V 261 | XP_016875235.1 | |
XM_017019747.1 | 695 | Missense Mutation | GCC,GTC | A,V 261 | XP_016875236.1 | |
XM_017019748.1 | 695 | Missense Mutation | GCC,GTC | A,V 128 | XP_016875237.1 | |
XM_017019749.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875238.1 | |
XM_017019750.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875239.1 | |
XM_017019751.1 | 695 | Missense Mutation | GCC,GTC | A,V 128 | XP_016875240.1 | |
XM_017019752.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875241.1 | |
XM_017019753.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875242.1 | |
XM_017019754.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875243.1 | |
XM_017019755.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875244.1 | |
XM_017019756.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875245.1 | |
XM_017019757.1 | 695 | Missense Mutation | GCC,GTC | A,V 113 | XP_016875246.1 |