Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTACATCAATGCAAATATCATCA[C/T]GGTAAGCTTTGCTTTTCACAGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPN11 - protein tyrosine phosphatase, non-receptor type 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002834.3 | 1312 | Missense Mutation | NP_002825.3 | |||
XM_006719526.2 | 1312 | Intron | XP_006719589.1 | |||
XM_011538613.2 | 1312 | Intron | XP_011536915.1 | |||
XM_017019722.1 | 1312 | Intron | XP_016875211.1 |