Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCATGGTTTCCATGTTAAGGAA[A/G]CGGACGTGCATCTTGGTCTCAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609073 MIM: 611585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXW8 - F-box and WD repeat domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TESC - tescalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168325.1 | 774 | Silent Mutation | CGC,CGT | R,R 176 | NP_001161797.1 | |
NM_017899.3 | 774 | Silent Mutation | CGC,CGT | R,R 203 | NP_060369.3 | |
XM_011538503.1 | 774 | Silent Mutation | CGC,CGT | R,R 191 | XP_011536805.1 | |
XM_017019532.1 | 774 | Silent Mutation | CGC,CGT | R,R 240 | XP_016875021.1 | |
XM_017019533.1 | 774 | UTR 3 | XP_016875022.1 |