Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTTTCACTGCCTGATAACCAGCC[A/G]TTTTTAATTTAGTGAAGAAAAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612157 | ||||||||||||||||||||
Literature Links: |
SENP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SENP1 - SUMO1/sentrin specific peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267594.1 | 2084 | Missense Mutation | ACG,ATG | T,M 503 | NP_001254523.1 | |
NM_001267595.1 | 2084 | Missense Mutation | ACG,ATG | T,M 503 | NP_001254524.1 | |
XM_006719361.3 | 2084 | Missense Mutation | ACG,ATG | T,M 535 | XP_006719424.1 | |
XM_006719362.2 | 2084 | Missense Mutation | ACG,ATG | T,M 304 | XP_006719425.1 | |
XM_011538244.2 | 2084 | Missense Mutation | ACG,ATG | T,M 522 | XP_011536546.1 | |
XM_011538245.2 | 2084 | Missense Mutation | ACG,ATG | T,M 519 | XP_011536547.1 | |
XM_017019229.1 | 2084 | Missense Mutation | ACG,ATG | T,M 566 | XP_016874718.1 | |
XM_017019230.1 | 2084 | Missense Mutation | ACG,ATG | T,M 553 | XP_016874719.1 | |
XM_017019231.1 | 2084 | Missense Mutation | ACG,ATG | T,M 534 | XP_016874720.1 | |
XM_017019232.1 | 2084 | Missense Mutation | ACG,ATG | T,M 534 | XP_016874721.1 | |
XM_017019233.1 | 2084 | Missense Mutation | ACG,ATG | T,M 496 | XP_016874722.1 | |
XM_017019234.1 | 2084 | Missense Mutation | ACG,ATG | T,M 496 | XP_016874723.1 | |
XM_017019235.1 | 2084 | Missense Mutation | ACG,ATG | T,M 496 | XP_016874724.1 | |
XM_017019236.1 | 2084 | Missense Mutation | ACG,ATG | T,M 490 | XP_016874725.1 | |
XM_017019237.1 | 2084 | Missense Mutation | ACG,ATG | T,M 490 | XP_016874726.1 | |
XM_017019238.1 | 2084 | Missense Mutation | ACG,ATG | T,M 304 | XP_016874727.1 | |
XM_017019239.1 | 2084 | Missense Mutation | ACG,ATG | T,M 304 | XP_016874728.1 | |
XM_017019240.1 | 2084 | Missense Mutation | ACG,ATG | T,M 291 | XP_016874729.1 |