Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTGCTGCCGTCACTCCCCAC[A/T]GGCCCTGAACAATCAAAAGTACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TUBGCP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUBGCP3 - tubulin gamma complex associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286277.1 | 3391 | Missense Mutation | CAG,CTG | Q,L 808 | NP_001273206.1 | |
NM_001286278.1 | 3391 | Intron | NP_001273207.1 | |||
NM_001286279.1 | 3391 | Intron | NP_001273208.1 | |||
NM_006322.5 | 3391 | Missense Mutation | CAG,CTG | Q,L 818 | NP_006313.1 | |
XM_005268293.1 | 3391 | Intron | XP_005268350.1 | |||
XM_011537462.2 | 3391 | Missense Mutation | CAG,CTG | Q,L 718 | XP_011535764.1 | |
XM_011537463.2 | 3391 | Missense Mutation | CAG,CTG | Q,L 589 | XP_011535765.1 | |
XM_011537464.2 | 3391 | Missense Mutation | CAG,CTG | Q,L 589 | XP_011535766.1 | |
XM_017020323.1 | 3391 | Missense Mutation | CAG,CTG | Q,L 589 | XP_016875812.1 |