Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATACACCAGTGCTAGAGTTCATC[A/G]TGTGTCAAAACTATAAGGGAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601184 MIM: 605898 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC3 - DnaJ heat shock protein family (Hsp40) member C3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UGGT2 - UDP-glucose glycoprotein glucosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020121.3 | 4745 | Silent Mutation | CAC,CAT | H,H 1513 | NP_064506.3 | |
XM_011521094.1 | 4745 | Intron | XP_011519396.1 | |||
XM_011521095.1 | 4745 | Intron | XP_011519397.1 | |||
XM_011521096.1 | 4745 | Intron | XP_011519398.1 | |||
XM_011521097.1 | 4745 | Silent Mutation | CAC,CAT | H,H 1540 | XP_011519399.1 | |
XM_011521098.1 | 4745 | Intron | XP_011519400.1 | |||
XM_011521099.2 | 4745 | Intron | XP_011519401.1 | |||
XM_011521100.2 | 4745 | Intron | XP_011519402.1 | |||
XM_011521102.2 | 4745 | Intron | XP_011519404.1 | |||
XM_011521103.2 | 4745 | Intron | XP_011519405.1 | |||
XM_017020662.1 | 4745 | Intron | XP_016876151.1 | |||
XM_017020663.1 | 4745 | Intron | XP_016876152.1 |