Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTGACGGTGGGGTTTAGCACC[C/G]CCCCAGTTGGCCCCAGAACCCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600297 | ||||||||||||||||||||
Literature Links: |
CDX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDX2 - caudal type homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265.4 | 1265 | Missense Mutation | GCG,GGG | A,G 305 | NP_001256.3 | |
XM_011534875.2 | 1265 | Missense Mutation | GCG,GGG | A,G 313 | XP_011533177.1 | |
XM_011534876.2 | 1265 | UTR 3 | XP_011533178.1 | |||
XM_011534877.2 | 1265 | UTR 3 | XP_011533179.1 | |||
XM_011534878.2 | 1265 | Missense Mutation | GCG,GGG | A,G 196 | XP_011533180.1 | |
XM_011534879.2 | 1265 | Missense Mutation | GCG,GGG | A,G 188 | XP_011533181.1 |
LINC00543 - long intergenic non-protein coding RNA 543 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |