Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCACCATGTTCCTGACTGCGC[A/T]CCTCTGGCGCGGCCGCATTCCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL57 - mitochondrial ribosomal protein L57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024026.4 | 141 | Missense Mutation | CAC,CTC | H,L 6 | NP_076931.1 | |
XM_017020740.1 | 141 | Missense Mutation | CAC,CTC | H,L 6 | XP_016876229.1 |
SKA3 - spindle and kinetochore associated complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166017.1 | 141 | Intron | NP_001159489.1 | |||
NM_145061.5 | 141 | Intron | NP_659498.4 | |||
XM_005266288.3 | 141 | Intron | XP_005266345.1 | |||
XM_011534994.1 | 141 | Intron | XP_011533296.1 |