Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTGCCCCGCGTCAGGTACTTGA[C/T]GGTTTCCGATGTTTTTCTGAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607587 | ||||||||||||||||||||
Literature Links: |
EEF1AKMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEF1AKMT1 - eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318939.1 | 590 | Missense Mutation | ATC,GTC | I,V 157 | NP_001305868.1 | |
NM_174928.2 | 590 | Missense Mutation | ATC,GTC | I,V 157 | NP_777588.1 | |
XM_017020432.1 | 590 | Missense Mutation | ATC,GTC | I,V 157 | XP_016875921.1 |
IL17D - interleukin 17D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |