Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGAAGCTGCTTCTCGAGTAAATA[A/G]TTTGAACCCACACTGTGTGTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG5 - ALG5, dolichyl-phosphate beta-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142364.1 | 785 | Silent Mutation | CTA,TTA | L,L 210 | NP_001135836.1 | |
NM_013338.4 | 785 | Silent Mutation | CTA,TTA | L,L 240 | NP_037470.1 | |
XM_017020552.1 | 785 | Silent Mutation | CTA,TTA | L,L 177 | XP_016876041.1 |