Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCAAGGGATTCAGCCTGGCGG[A/C]CATCCCCTACCAGGAGATCGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPC6 - glypican 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005708.3 | 769 | Missense Mutation | GAC,GCC | D,A 46 | NP_005699.1 | |
XM_011521044.2 | 769 | Intron | XP_011519346.1 | |||
XM_017020298.1 | 769 | Intron | XP_016875787.1 | |||
XM_017020299.1 | 769 | Intron | XP_016875788.1 | |||
XM_017020300.1 | 769 | Intron | XP_016875789.1 | |||
XM_017020301.1 | 769 | Intron | XP_016875790.1 | |||
XM_017020302.1 | 769 | Intron | XP_016875791.1 |