Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAATTTGCAGCTTCCTTCAAAAG[C/T]GTAAGTAATATCCCTTACCTACTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LIN52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIN52 - lin-52 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024674.2 | 276 | Silent Mutation | AGC,AGT | S,S 48 | NP_001019845.1 | |
XM_011537320.2 | 276 | Silent Mutation | AGC,AGT | S,S 80 | XP_011535622.1 | |
XM_011537321.2 | 276 | Silent Mutation | AGC,AGT | S,S 80 | XP_011535623.1 | |
XM_011537322.2 | 276 | Silent Mutation | AGC,AGT | S,S 48 | XP_011535624.2 | |
XM_017021763.1 | 276 | Silent Mutation | AGC,AGT | S,S 46 | XP_016877252.1 | |
XM_017021764.1 | 276 | Silent Mutation | AGC,AGT | S,S 48 | XP_016877253.1 |