Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCCACACCAGGACTCTATCCTA[C/G]TCCCAGTAATCCTTTCCAAGTGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MAPK1IP1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPK1IP1L - mitogen-activated protein kinase 1 interacting protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144578.3 | 1667 | Missense Mutation | ACT,AGT | T,S 220 | NP_653179.1 | |
XM_011537362.2 | 1667 | Missense Mutation | ACT,AGT | T,S 233 | XP_011535664.2 | |
XM_017021789.1 | 1667 | Missense Mutation | ACT,AGT | T,S 220 | XP_016877278.1 |