Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCATCATCTTCACTGTCCTGGAA[A/C]CTTGGCAACTAGAAAACAAGTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCM - Fanconi anemia complementation group M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIS18BP1 - MIS18 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018353.4 | 3499 | Missense Mutation | AGG,AGT | R,S 1004 | NP_060823.3 | |
XM_005267833.4 | 3499 | Missense Mutation | AGG,AGT | R,S 1004 | XP_005267890.1 | |
XM_011536923.1 | 3499 | Missense Mutation | AGG,AGT | R,S 616 | XP_011535225.1 | |
XM_017021425.1 | 3499 | Missense Mutation | AGG,AGT | R,S 1004 | XP_016876914.1 |