Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAATTATATTTAAGGCAATGGG[C/T]TTTTTGATGATGTCATAGTAGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605680 | ||||||||||||||||||||
Literature Links: |
BAZ1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAZ1A - bromodomain adjacent to zinc finger domain 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013448.2 | 3111 | Silent Mutation | AAA,AAG | K,K 1472 | NP_038476.2 | |
NM_182648.1 | 3111 | Silent Mutation | AAA,AAG | K,K 1440 | NP_872589.1 | |
XM_011536374.1 | 3111 | Silent Mutation | AAA,AAG | K,K 1379 | XP_011534676.1 | |
XM_011536376.2 | 3111 | Silent Mutation | AAA,AAG | K,K 995 | XP_011534678.2 | |
XM_017020942.1 | 3111 | Silent Mutation | AAA,AAG | K,K 963 | XP_016876431.1 |