Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGAGCCAACAGCCGTTCCCAGAA[C/G]CTGCAATCCGCTGCGCCCCCAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611137 MIM: 600306 | ||||||||||||||||||||
Literature Links: |
PSMB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMB11 - proteasome subunit beta 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMB5 - proteasome subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130725.1 | 595 | Missense Mutation | AGC,AGG | S,R 9 | NP_001124197.1 | |
NM_001144932.2 | 595 | Missense Mutation | AGC,AGG | S,R 112 | NP_001138404.1 | |
NM_002797.4 | 595 | Missense Mutation | AGC,AGG | S,R 112 | NP_002788.1 | |
XM_005267871.2 | 595 | Missense Mutation | AGC,AGG | S,R 112 | XP_005267928.1 |