Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAAGGCAGTGCCAGAAAAATTAC[A/G]ACCACTGCTGGATAGTCTGGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACIN1 - apoptotic chromatin condensation inducer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C14orf119 - chromosome 14 open reading frame 119 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017924.3 | 359 | Missense Mutation | CAA,CGA | Q,R 71 | NP_060394.1 | |
XM_017021390.1 | 359 | Missense Mutation | CAA,CGA | Q,R 71 | XP_016876879.1 |
LOC100128908 - leishmanolysin homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |