Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCCACATAAAGCCCCTTCCG[C/T]TTTCGTACTGCGTTCATGTACTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EFL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFL1 - elongation factor like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040610.2 | 3304 | Silent Mutation | AAA,AAG | K,K 1045 | NP_001035700.1 | |
NM_001322844.1 | 3304 | Silent Mutation | AAA,AAG | K,K 833 | NP_001309773.1 | |
NM_001322845.1 | 3304 | Silent Mutation | AAA,AAG | K,K 1096 | NP_001309774.1 | |
NM_024580.5 | 3304 | Silent Mutation | AAA,AAG | K,K 1096 | NP_078856.4 | |
XM_011522000.1 | 3304 | Silent Mutation | AAA,AAG | K,K 833 | XP_011520302.1 |