Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTGCGCCATCGACTGTCTCAA[C/T]CTGGCCACGGGCCAGTGGACGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KBTBD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KBTBD13 - kelch repeat and BTB domain containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101362.2 | 1119 | Silent Mutation | AAC,AAT | N,N 373 | NP_001094832.1 |
RASL12 - RAS like family 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |