Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGGCGTTACTGCAGGTCTGGG[A/C]GGCCGAATCGGCGCTGCGTGGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603484 MIM: 611219 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRC1 - protein regulator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRC1-AS1 - PRC1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCCD1 - RCC1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017919.1 | 495 | Missense Mutation | GAG,GCG | E,A 100 | NP_001017919.1 | |
NM_033544.2 | 495 | Missense Mutation | GAG,GCG | E,A 100 | NP_291022.2 |
UNC45A - unc-45 myosin chaperone A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |