Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGCCATGGGTGGCGCCTCCCC[C/T]TACTGCCTGCCCAGGGCTCTGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147650 | ||||||||||||||||||||
Literature Links: |
IDH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDH2 - isocitrate dehydrogenase (NADP(+)) 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289910.1 | 1265 | Silent Mutation | TAA,TAG | *,* 401 | NP_001276839.1 | |
NM_001290114.1 | 1265 | Silent Mutation | TAA,TAG | *,* 323 | NP_001277043.1 | |
NM_002168.3 | 1265 | Silent Mutation | TAA,TAG | *,* 453 | NP_002159.2 |
ZNF710 - zinc finger protein 710 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |