Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACCTGAGGAAGTCAGGGGCCT[G/T]GGCAATCATGTCCTCGAAGTCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIB2 - calcium and integrin binding family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271888.1 | 668 | Missense Mutation | AAG,CAG | K,Q 132 | NP_001258817.1 | |
NM_001271889.1 | 668 | Missense Mutation | AAG,CAG | K,Q 126 | NP_001258818.1 | |
NM_001301224.1 | 668 | Missense Mutation | AAG,CAG | K,Q 180 | NP_001288153.1 | |
NM_006383.3 | 668 | Missense Mutation | AAG,CAG | K,Q 175 | NP_006374.1 | |
XM_005254126.3 | 668 | Missense Mutation | AAG,CAG | K,Q 175 | XP_005254183.1 | |
XM_006720374.2 | 668 | Missense Mutation | AAG,CAG | K,Q 132 | XP_006720437.1 | |
XM_011521161.2 | 668 | Missense Mutation | AAG,CAG | K,Q 132 | XP_011519463.1 |
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |