Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCGGGGCCAGGTTCACCCAGC[C/T]GGGCCCGCGGGGCAGCAGGTGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT12 - septin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001154458.2 | 1240 | Missense Mutation | AGC,GGC | S,G 280 | NP_001147930.1 | |
NM_144605.4 | 1240 | Missense Mutation | AGC,GGC | S,G 326 | NP_653206.2 | |
XM_006720846.2 | 1240 | Missense Mutation | AGC,GGC | S,G 326 | XP_006720909.1 | |
XM_011522379.2 | 1240 | Missense Mutation | AGC,GGC | S,G 262 | XP_011520681.1 | |
XM_017022938.1 | 1240 | Missense Mutation | AGC,GGC | S,G 332 | XP_016878427.1 |