Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCAGAGGAGGGGGCTGGAAAT[A/G]CCACCCTGAGCCCTAGAATGGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613199 | ||||||||||||||||||||
Literature Links: |
LOC107987231 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107987231 - uncharacterized LOC107987231 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAOK2 - TAO kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM219 - transmembrane protein 219 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083613.1 | 641 | Missense Mutation | ACC,GCC | T,A 168 | NP_001077082.1 | |
NM_194280.3 | 641 | Missense Mutation | ACC,GCC | T,A 168 | NP_919256.1 | |
XM_005255123.1 | 641 | Missense Mutation | ACC,GCC | T,A 168 | XP_005255180.1 |