Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCCAGGGCGGATCCCTTGCAG[C/G]ACACCACTGGGGCCCAGCACCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLYR1 - glyoxylate reductase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308096.1 | 1206 | Intron | NP_001295025.1 | |||
NM_001324096.1 | 1206 | Intron | NP_001311025.1 | |||
NM_001324097.1 | 1206 | Intron | NP_001311026.1 | |||
NM_001324098.1 | 1206 | Intron | NP_001311027.1 | |||
NM_032569.3 | 1206 | Intron | NP_115958.2 | |||
XM_005255639.4 | 1206 | Intron | XP_005255696.2 | |||
XM_011522716.2 | 1206 | Silent Mutation | GTC,GTG | V,V 342 | XP_011521018.1 | |
XM_011522717.1 | 1206 | Intron | XP_011521019.1 | |||
XM_017023786.1 | 1206 | Intron | XP_016879275.1 | |||
XM_017023787.1 | 1206 | Intron | XP_016879276.1 | |||
XM_017023788.1 | 1206 | Intron | XP_016879277.1 | |||
XM_017023789.1 | 1206 | Intron | XP_016879278.1 |