Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTAAATTCATCAGTCTTTCCCT[C/T]ATGTGGGTTCTCAGGGCAATACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZKSCAN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZKSCAN2 - zinc finger with KRAB and SCAN domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012981.4 | 3276 | Missense Mutation | AAG,GAG | K,E 957 | NP_001012999.3 | |
XM_017023200.1 | 3276 | Missense Mutation | AAG,GAG | K,E 753 | XP_016878689.1 |