Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCGTGAGGGACGACGGCAGCTCC[A/T]TGTCCTTCTACGTGCGGCCCAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601421 MIM: 605061 | ||||||||||||||||||||
Literature Links: |
KARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KARS - lysyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130089.1 | Intron | NP_001123561.1 | ||||
NM_005548.2 | Intron | NP_005539.1 | ||||
XM_017023217.1 | Intron | XP_016878706.1 |
TERF2IP - TERF2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018975.3 | Intron | NP_061848.2 |