Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGATGTAGACCCCGGGCCTGTTG[T/C]GCTCGGCACAGCCCTCGCCCCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609343 | ||||||||||||||||||||
Literature Links: |
PRSS22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS22 - protease, serine 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022119.3 | 872 | Missense Mutation | CAC,CGC | H,R 269 | NP_071402.1 | |
XM_005255473.3 | 872 | Missense Mutation | CAC,CGC | H,R 269 | XP_005255530.1 |