Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATCCACCGATGGGCTTCCAGGG[C/T]GGTCTTGCGCTTGAAGCCTTTCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF785 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF785 - zinc finger protein 785 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152458.6 | 1526 | Missense Mutation | ACC,GCC | T,A 354 | NP_689671.2 | |
XM_011545752.2 | 1526 | Missense Mutation | ACC,GCC | T,A 367 | XP_011544054.1 | |
XM_011545753.2 | 1526 | Missense Mutation | ACC,GCC | T,A 367 | XP_011544055.1 | |
XM_017022967.1 | 1526 | Missense Mutation | ACC,GCC | T,A 354 | XP_016878456.1 |