Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGTCGCTATTCATAATGGCTG[G/T]TGCAATTCCTTCATGAATAGATCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602212 | ||||||||||||||||||||
Literature Links: |
LOC100507577 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507577 - uncharacterized LOC100507577 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LONP2 - lon peptidase 2, peroxisomal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIAH1 - siah E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006610.1 | 2378 | Missense Mutation | ACA,CCA | T,P 280 | NP_001006611.1 | |
NM_003031.3 | 2378 | Missense Mutation | ACA,CCA | T,P 249 | NP_003022.3 | |
XM_006721246.1 | 2378 | Missense Mutation | ACA,CCA | T,P 249 | XP_006721309.1 | |
XM_011523279.1 | 2378 | Missense Mutation | ACA,CCA | T,P 249 | XP_011521581.1 |