Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGCCGTGGCGGAAGGCCAGGG[A/C]GCGGAAGCTGGGCAGGCCCGGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HS3ST6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS3ST6 - heparan sulfate-glucosamine 3-sulfotransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009606.3 | 842 | Missense Mutation | NP_001009606.3 | |||
XM_011522608.2 | 842 | Missense Mutation | XP_011520910.1 | |||
XM_011522609.2 | 842 | Missense Mutation | XP_011520911.1 |