Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGCACAAGCCCCTGAACCATAT[A/G]TTGTTATCACTGGATTTGAAGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616074 MIM: 607884 MIM: 188250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKLF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKLF - chemokine-like factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040138.2 | 289 | Missense Mutation | ATT,GTT | I,V 46 | NP_001035228.1 | |
NM_016326.3 | 289 | Intron | NP_057410.1 | |||
NM_016951.3 | 289 | Missense Mutation | ATT,GTT | I,V 46 | NP_058647.1 | |
NM_181640.2 | 289 | Intron | NP_857591.1 | |||
NM_181641.2 | 289 | Missense Mutation | ATT,GTT | I,V 46 | NP_857592.1 |
CKLF-CMTM1 - CKLF-CMTM1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202509.1 | 289 | Intron | NP_001189438.1 | |||
NM_001204098.1 | 289 | Intron | NP_001191027.1 | |||
NM_001204099.1 | 289 | Missense Mutation | ATT,GTT | I,V 46 | NP_001191028.1 |
CMTM1 - CKLF like MARVEL transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TK2 - thymidine kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |