Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGGTTGGGCCGTGTTCTTAGC[A/G]AGCAGAAGCCTTGGCCAGGGTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPRY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSPRY1 - ring finger and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305163.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | NP_001292092.1 | |
NM_001305164.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | NP_001292093.1 | |
NM_001305182.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | NP_001292111.1 | |
NM_133368.2 | 356 | Silent Mutation | GCA,GCG | A,A 11 | NP_588609.1 | |
XM_005256220.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | XP_005256277.1 | |
XM_011523427.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | XP_011521729.1 | |
XM_011523428.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | XP_011521730.1 | |
XM_011523430.1 | 356 | Silent Mutation | GCA,GCG | A,A 11 | XP_011521732.1 | |
XM_017023844.1 | 356 | UTR 5 | XP_016879333.1 |