Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGCGCACAGAGCTCTCGCCCA[C/T]GGCAGAGGCGCCCTCGGGCAGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164010 MIM: 179780 | ||||||||||||||||||||
Literature Links: |
CHMP1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP1A - charged multivesicular body protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083314.3 | 720 | Missense Mutation | CAT,CGT | H,R 168 | NP_001076783.1 | |
NM_002768.4 | 720 | Missense Mutation | ATG,GTG | M,V 175 | NP_002759.2 | |
XM_011523098.1 | 720 | Intron | XP_011521400.1 |
DPEP1 - dipeptidase 1 (renal) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |