Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609377 MIM: 607484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACD - adrenocortical dysplasia homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf86 - chromosome 16 open reading frame 86 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012984.2 | 659 | Missense Mutation | CCC,CTC | P,L 165 | NP_001013002.2 | |
XM_005255952.4 | 659 | Missense Mutation | CCC,CTC | P,L 165 | XP_005256009.1 |
ENKD1 - enkurin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFOD2 - glucose-fructose oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARD6A - par-6 family cell polarity regulator alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |