Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGTGGGTGCCGGAGCAGAGCAGC[A/G]CCGTGGTCCTGGCGGTCCTGCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987233 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987233 - uncharacterized LOC107987233 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NHLRC4 - NHL repeat containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGQ - phosphatidylinositol glycan anchor biosynthesis class Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004204.3 | 226 | Missense Mutation | ACC,GCC | T,A 30 | NP_004195.2 | |
NM_148920.2 | 226 | Missense Mutation | ACC,GCC | T,A 30 | NP_683721.1 |
PRR35 - proline rich 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |