Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAGAGCGAGACAAGGCTCTGCA[A/G]GAGCTTCGGCAAGAGCTGAACACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605821 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHSP - alpha hemoglobin stabilizing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318221.1 | 319 | Silent Mutation | CAA,CAG | Q,Q 68 | NP_001305150.1 | |
NM_001318222.1 | 319 | Silent Mutation | CAA,CAG | Q,Q 68 | NP_001305151.1 | |
NM_016633.3 | 319 | Silent Mutation | CAA,CAG | Q,Q 68 | NP_057717.1 |